Sodium peroxodisulfate ≥99.0% (by iodometry), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.06609.0500
1.06609.1000
1.06609.9025
EM1.06609.0500CS
1019
USD
EM1.06609.0500
EM1.06609.1000
EM1.06609.9025
Sodium peroxodisulfate ≥99.0% (by iodometry), EMSURE® for analysis, Supelco®
Sodium peroxodisulphate
In North America 1.06609.0500 replaces, but with a far superior specification to SX0703-1
Formula:
Na₂S₂O₈ Melting Pt: 100 °C Density: 1.2 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003501 CAS Number: 7775-27-1 EINECS: 231-892-1 UN: 1505 ADR: 5.1,III Merck Index: 13,08729 |
Specification Test Results
Assay (iodometric) | ≥ 99.0 % |
Identity | passes test |
Chloride (Cl) | ≤ 0.01 % |
Heavy metals (as Pb) | ≤ 0.003 % |
NH₄ (Ammonium) | ≤ 0.05 % |
Cr (Chromium) | ≤ 0.001 % |
Cu (Copper) | ≤ 0.001 % |
Fe (Iron) | ≤ 0.0005 % |
Mn (Manganese) | ≤ 0.0001 % |
Pb (Lead) | ≤ 0.001 % |
Loss on drying (105 °C; 3 hours) | ≤ 0.05 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...