Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
Supplier: MilliporeSigma
1.06587.1000
EM1.06587.1000CS
511
USD
EM1.06587.1000
Di-Sodium Hydrogen Phosphate Solution (Buffer Stock Solution), Certipur®
Buffers
pH Calibration Buffers
Buffer stock solution 1/15 mol/l
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...