Sodium perborate tetrahydrate ≥96.0% (by manganometry), pure, Supelco®

Supplier: MilliporeSigma
Danger

1.06560.1000 1.06560.9050
EM1.06560.1000CS 450 USD
EM1.06560.1000 EM1.06560.9050
Sodium perborate tetrahydrate ≥96.0% (by manganometry), pure, Supelco®
Sodium perborate tetrahydrate

EMD # 1.06560.9050 replaces and is identical to EMD # 1.06560.9057.

Formula: NaH₂BO₄·4H₂O
Melting Pt: 60 °C (dec.)
Density: 1.73 g/cm3
Storage Temperature: Ambient
MDL Number: MFCD00149231
CAS Number: 13517-20-9
EINECS: 239-172-9

Order Now

Specification Test Results

Solubility 23 g/l (20 °C)
Melting Point 60 °C (decomposition)
Molar Mass 153.88 g/mol
Bulk Density 800 kg/m3
Density 1.73 g/cm3
pH 10 (15 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR