Sodium nitrate, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.06546.0050
EM1.06546.0050CS 21847 USD
EM1.06546.0050
Sodium nitrate, Suprapur® for trace metal analysis, Supelco®
Sodium nitrate
Formula: NaNO₃
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 13,08720

Order Now

Specification Test Results

Solubility 874 g/l (20 °C)
Melting Point 308 °C
Molar Mass 84.99 g/mol
Bulk Density 1200 kg/m3
Density 2.26 g/cm3 (20 °C)
pH 5.5 - 8.0 (50 g/l, H2O, 20 °C)
Hazard Class 5.1
Hazard Class Description Oxidizer


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR