Sodium iodide, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Hydriodic acid sodium salt, Sodium monoiodide, Ioduril

1.06523.0100 1.06523.0250 1.06523.1000
EM1.06523.0100CS 20050 USD
EM1.06523.0100 EM1.06523.0250 EM1.06523.1000
Sodium iodide, EMSURE® for analysis, Supelco®
Sodium iodide
Formula: NaI
MW: 149.89 g/mol
Boiling Pt: 1300
Melting Pt: 662
Density: 3.67 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003532
CAS Number: 7681-82-5
EINECS: 231-679-3
UN: 3077
ADR: 9 III
Merck Index: 13,08705

Order Now

Specification Test Results

Solubility 1793 g/l (20 °C)
Melting Point 662 °C
Molar Mass 149.89 g/mol
Bulk Density 1500 - 2000 kg/m3
Boiling Point 1304 °C (1013 hPa)
Vapor Pressure 1.3 hPa (767 °C)
Density 3.67 g/cm3 (20 °C)
pH 6 - 9 (50 g/l, H2O, 20 °C)
Hazard Class 9
Hazard Class Description Miscellaneous Hazardous Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR