Sodium molybdate(VI) dihydrate, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
1.06521.0100
1.06521.0250
1.06521.1000
EM1.06521.0100CS
12343
USD
EM1.06521.0100
EM1.06521.0250
EM1.06521.1000
Sodium molybdate(VI) dihydrate, EMSURE® for analysis, Supelco®
Sodium molybdate(VI) dihydrate
Formula:
Na₂MoO₄·2H₂O Melting Pt: 687 °C Density: 3.28 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00149170 CAS Number: 10102-40-6 EINECS: 231-551-7 Merck Index: 13,08718 |
Specification Test Results
Solubility | 840 g/l (20 °C) |
Melting Point | 687 °C Elimination of water of crystallisation |
Molar Mass | 241.95 g/mol |
Bulk Density | 1000 - 1400 kg/m3 |
Density | 3.28 g/cm3 (20 °C) (for the dihydrate) |
pH | 7.9 - 10.3 (50 g/l, H2O, 20 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...