Sodium hydroxide ≥99% (NaOH basis by acidimetry), pellets, EMSURE® ISO ISO for LC-MS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
M-Clarity™ quality level = MQ300
1.06498.1000
1.06498.0500
1.06498.5000
1.06498.9025
1.06498.9050
EM1.06498.1000EA
154
USD
EM1.06498.1000
EM1.06498.0500
EM1.06498.5000
EM1.06498.9025
EM1.06498.9050
Sodium hydroxide ≥99% (NaOH basis by acidimetry), pellets, EMSURE® ISO ISO for LC-MS, Supelco®
Sodium hydroxide
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 318 °C (lit.) Density: 2.13 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
Assay (acidimetric, NaOH) | 99% min |
Carbonate (as Na₂CO₃) | 1.0% max |
Chloride (Cl) | 0.012% max |
Phosphate (PO₄) | 0.0005% max |
Silicate (SiO₂) | 0.001% max |
Sulfate (SO₄) | 0.010% max |
Total nitrogen (N) | 0.0003% max |
Heavy metals (as Pb) | 0.0005% max |
Al (Aluminium) | 0.0005% max |
As (Aresnic) | 0.0001% max |
Ca (Calcium) | 0.0005% max |
Cu (Copper) | 0.0002% max |
Fe (Iron) | 0.0005% max |
K (Pottassium) | 0.05% max |
Mg (Magnesium) | 0.0005% max |
Ni (Nickel) | 0.00025% max |
Pb (Lead) | 0.0005% max |
Zn (Zinc) | 0.001% max |
. | . |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...