Sodium hydroxide 99-100% (total alkilinity calculated as NaOH), pellets, EMSURE® ACS, Reag. Ph Eur Reag. Ph. Eur., ACS for LC-MS (max. 0.02% K), Supelco®
Supplier: MilliporeSigma
|
Danger
|
Synonyms:
Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye
M-Clarity™ quality level = MQ300
1.06469.9050
1.06469.1000
1.06469.5000
1.06469.9012
CASX0590-7EA
486.62
USD
CASX0590-7
EM1.06469.9050
EM1.06469.1000
EM1.06469.5000
EM1.06469.9012
Sodium hydroxide 99-100% (total alkilinity calculated as NaOH), pellets, EMSURE® ACS, Reag. Ph Eur Reag. Ph. Eur., ACS for LC-MS (max. 0.02% K), Supelco®
Sodium hydroxide
Pellets.
In North America 1.06469.9050 replaces and is identical to SX0590-20
Formula:
NaOH Boiling Pt: 1390 °C (1013 hPa) Melting Pt: 318 °C (lit.) Density: 2.13 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00003548 CAS Number: 1310-73-2 EINECS: 215-185-5 UN: 1823 ADR: 8,II REACH: 01-2119457892-27 Merck Index: 14,08627 |
Specification Test Results
Assay (acidimetric,NaOH) | 99 - 100 % |
Assay (total alkalinity calc. as NaOH) | 99 - 100 % |
Identity | passes test |
Appearance of solution | passes test |
Carbonate (as Na₂CO₃) | ≤ 0.5 % |
Chloride (Cl) | ≤ 0.0005 % |
Phosphate (PO₄) | ≤ 0.0005 % |
Silicate (SiO₂) | ≤ 0.001 % |
Sulfate (SO₄) | ≤ 0.0005 % |
Total nitrogen (N) | ≤ 0.0003 % |
Heavy metals (as Ag) | ≤ 0.0005 % |
Heavy metals (as Pb) | ≤ 0.0005 % |
Al (Aluminium) | ≤ 0.0005 % |
As (Arsenic) | ≤ 0.0001 % |
Ca (Calcium) | ≤ 0.0005 % |
Cu (Copper) | ≤ 0.0002 % |
Fe (Iron) | ≤ 0.0005 % |
Hg (Mercury) | ≤ 0.00001 % |
K (Potassium) | ≤ 0.020 % |
Mg (Magnesium) | ≤ 0.0005 % |
Ni (Nickel) | ≤ 0.0005 % |
Pb (Lead) | ≤ 0.0005 % |
Zn (Zinc) | ≤ 0.001 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...