Sodium fluoride, Suprapur® Trace_metal, Supelco®

Supplier: MilliporeSigma
Danger

1.06450.0025
EM1.06450.0025CS 28295 USD
EM1.06450.0025
Sodium fluoride, Suprapur® Trace_metal, Supelco®
Sodium fluoride
Formula: NaF
MW: 41.99 g/mol
Boiling Pt: 1700
Melting Pt: 993
Density: 2.8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003524
CAS Number: 7681-49-4
EINECS: 231-667-8
UN: 1690
ADR: 6.1 III
Merck Index: 13,08691
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 42 g/l (20 °C)
Melting Point 993 °C
Molar Mass 41.98 g/mol
Bulk Density 800 kg/m3
Boiling Point 1704 °C
Vapor Pressure 1 hPa (1077 °C)
Density 2.8 g/cm3 (20 °C)
pH 10.2 (40 g/l, H2O, 20 °C)
Hazard Class 6.1
Hazard Class Description Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR