tri-Sodium citrate dihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% anhydrous basis (perchloric acid titration)], Supelco®

Supplier: MilliporeSigma

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

M-Clarity™ quality level = MQ300

1.06448.9025
EM1.06448.9025EA 1444 USD
EM1.06448.9025
tri-Sodium citrate dihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% anhydrous basis (perchloric acid titration)], Supelco®
tri-Sodium citrate dihydrate
Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: >300 °C (lit.)
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR