tri-Sodium citrate dihydrate ≥99.0%, crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®

Supplier: MilliporeSigma

Synonyms: Sodium Citrate Dihydrate, Sodium citrate tribasic dihydrate, Trisodium citrate, Sodium Citrate, Dihydrate, Sodium citrate dihydrate, Citric acid tridosium salt dihydrate, Sodium 2-hydroxypropane-1,2,3-tricarboxylate dihydrate, Citric acid trisodium salt dihydrate

1.06432.9029
EM1.06432.9029EA 1660.94 USD
EM1.06432.9029
tri-Sodium citrate dihydrate ≥99.0%, crystals, EMPROVE® ESSENTIAL Ph. Eur., BP, JP, USP, E331, SAFC®
tri-Sodium citrate dihydrate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).


Certifications: Corresponds to Ph. Eur., BP, JP, USP, E 331
Conforms to the purity criteria on food additives according to the current European Commission Regulation.

Formula: Na₃C₆H₅O₇·2H₂O
MW: 294.1 g/mol
Melting Pt: 150 °C (anhydr.)
Density: 1.76 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150031
CAS Number: 6132-04-3
EINECS: 200-675-3
Merck Index: 13,08675
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e32147a215e8895f4b6db5aab1b99da667b795ac&vid=b929cda9d6f2eed18a8fefe57aa02b561af7c97f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (Perchloric acid titration, calc. on anhydrous substance (Ph Eur)) 99.0 - 101.0 %
Assay (Perchloric acid titration, previously dried substance) (JP/USP) 99.0 - 100.5 %
Identity (Na) passes test
Identity (Citrate) passes test
Identity (reaction upon ignition) passes test
Appearance white to almost white crystals
Appearance of solution (100 g/l, CO₂-free water) clear and colorless
Acidity or alkalinity passes test
pH (50 g/l CO₂-free water) 7.5 - 8.5
Chloride (Cl) ≤ 50 ppm
Sulfate (SO₄) ≤ 150 ppm
Heavy metals (as Pb) ≤ 5 ppm
Al (Aluminium) ≤ 5 ppm
As (Arsenic) ≤ 1 ppm
Hg (Mercury) ≤ 1 ppm
Pb (Lead) ≤ 1 ppm
Oxalate (as C₂H₂O₄) ≤ 100 ppm
Tartrate (C₄H₄O₆) passes test
Residual solvents (ICH (Q3C)) excluded by manufacturing process
Readily carbonisable substance passes test
Water (according to Karl Fischer) 11.0 - 13.0 %
Loss on drying (180 °C, 18 h) 10.0 - 13.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR