Sodium dihydrogen phosphate, anhydrous, Suprapur® for trace metal analysis, Supelco®

Supplier: MilliporeSigma

1.06370.0050
EM1.06370.0050CS 29376 USD
EM1.06370.0050
Sodium dihydrogen phosphate, anhydrous, Suprapur® for trace metal analysis, Supelco®
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Specification Test Results

Solubility 850 g/l (20 °C)
Molar Mass 119.98 g/mol
Bulk Density 940 kg/m3
pH 4.5 (10 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR