Sodium hydrogen sulfate monohydrate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06352.0500
EM1.06352.0500CS 1997 USD
EM1.06352.0500
Sodium hydrogen sulfate monohydrate, EMSURE® for analysis, Supelco®
Sodium hydrogen sulphate monohydrate
Formula: NaHSO₄·H₂O
Melting Pt: 183 °C
Density: 2.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149210
CAS Number: 10034-88-5
EINECS: 231-665-7
UN: 3260
ADR: 8,III
Merck Index: 13,08658

Order Now

Specification Test Results

Solubility 1080 g/l (20 °C)
Melting Point 183 °C (decomposition)
Molar Mass 138.07 g/mol
Bulk Density 900 - 970 kg/m3
Density 2.12 g/cm3 (20 °C)
pH 1 (50 g/l, H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR