Sodium benzoate 99.0-101.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211

Supplier: MilliporeSigma
Warning

Synonyms: Benzoic acid sodium salt

1.06290.9525
EM1.06290.9525EA 27524.69 USD
EM1.06290.9525
Sodium benzoate 99.0-101.0% (by HPLC on dried basis), EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E211
Sodium benzoate

Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1 to 3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated.

Formula: C₆H₅COONa
MW: 144.11 g/mol
Melting Pt: 436 °C
Density: 1.50 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012463
CAS Number: 532-32-1
EINECS: 208-534-8

Order Now

Specification Test Results

Assay (Acidimetric, Calculated on Anhydrous Substance) 99.0 - 100.5%
Assay (HPLC, Calculated on Anhydrous Substance) 99.0 - 101.0%
Identity Passes Test
Appearance of Solution (100 g/l, Water) Passes Test
Acidity or Alkalinity Passes Test
Chloride (Cl) ≤0.02%
Sulfate (SO₄) ≤0.01%
Total chlorine ≤0.03%
Heavy metals (as Pb) ≤10 ppm
As (Arsenic) ≤1 ppm
Cd (Cadmium) ≤10 ppm
Cu (Copper) ≤10 ppm
Hg (Mercury) ≤1 ppm
Pb (Lead) ≤2 ppm
Zn (Zinc) ≤10 ppm
Benzene ≤2 ppm
Toluene Excluded by Production Process
Multinuclear acids Passes Test
Oxidizable matter Passes Test
Water (According to Karl Fischer) ≤1.5%
Corresponds to Ph. Eur., BP, NF, FCC, E211

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR