Sodium metaarsenite 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Sodium dioxoarsenate

M-Clarity™ quality level = MQ100

1.06277.1000
EM1.06277.1000CS 438 USD
EM1.06277.1000
Sodium metaarsenite 0.05 mol/l (0.1 N) in aqueous solution, Titripur® Reag. USP volumetric solution, Supelco®
Sodium metaarsenite
Formula: NaAsO₂
MW: 129.91
Melting Pt: 615
Density: 1.87
Storage Temperature: Ambient
MDL Number: MFCD00003472
CAS Number: 7784-46-5
EINECS: 232-070-5
UN: 1686
ADR: 6.1 II
Merck Index: 13,08651

Order Now

Specification Test Results

Form liquid
Amount-of-substance concentration 0.04975 - 0.05025
Measurement uncertainty +/- 0.00015
Traceability NIST SRM
The concentration is determined by volumetric titration and refers to 20°C. Toxic Materials


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR