Sodium acetate ≥99.0% (by perchloric acid titration), Supelco®

Supplier: MilliporeSigma

Synonyms: Acetic acid sodium salt

M-Clarity™ quality level = MQ300

1.06268.9025
EM1.06268.9025EA 3910 USD
EM1.06268.9025
Sodium acetate ≥99.0% (by perchloric acid titration), Supelco®
Sodium acetate
Formula: H₃CCOONa
MW: 82.03 g/mol
Boiling Pt: >400 °C (dec.)
Melting Pt: 324 °C (dec.)
Density: 1.52 g/cm3 (20 °C)
Flash Pt: >250 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00012459
CAS Number: 127-09-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR