Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% dry basis (perchloric acid titration)], EMSURE® for analysis indifferent to potassium permanganate, Supelco®
Supplier: MilliporeSigma
Synonyms:
Acetic acid sodium salt trihydrate
1.06267.0500
1.06267.1000
1.06267.5000
1.06267.9050
EM1.06267.0500CS
1284
USD
EM1.06267.0500
EM1.06267.1000
EM1.06267.5000
EM1.06267.9050
Sodium acetate trihydrate 99.0-101.0% (by perchloric acid titration) [99.0-101.0% dry basis (perchloric acid titration)], EMSURE® for analysis indifferent to potassium permanganate, Supelco®
Sodium acetate trihydrate
Formula:
H₃CCOONa·3H₂O Boiling Pt: 120 - 123 °C (1013 hPa) Melting Pt: 58 °C Density: 1.42 g/cm3 (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00071557 CAS Number: 6131-90-4 EINECS: 204-823-8 Merck Index: 13,08642 |
|
Specification Test Results
Solubility | 613 g/l (20 °C) |
Melting Point | 58 °C |
Molar Mass | 136.08 g/mol |
Bulk Density | 900 kg/m3 |
Boiling Point |
|
Vapor Pressure | (20 °C) |
Flash Point |
|
Density | 1.42 g/cm3 (20 °C) |
pH | 8 - 9.5 (10 g/l, H2O, 20 °C) |
Ignition Point | 607 °C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...