Sodium, paraffin protect rods, EMSURE® 2.5 cm for analysis, Supelco®

Supplier: MilliporeSigma
Danger

1.06260.0250
EM1.06260.0250CS 1356 USD
EM1.06260.0250
Sodium, paraffin protect rods, EMSURE® 2.5 cm for analysis, Supelco®
Sodium
Formula: Na
MDL Number: MFCD00085307
CAS Number: 7440-23-5
UN: 1428
ADR: 4.3,I

Order Now

Specification Test Results

Solubility (20 °C) explosion decomposition
Melting Point 97.8 °C
Molar Mass 22.99 g/mol
Boiling Point 889 °C (1013 hPa)
Vapor Pressure 1.6 hPa (400 °C)
Density 0.97 g/cm3 (20 °C)
Ignition Point 115 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR