α-Naphtholphthalein indicator, Supelco®

Supplier: MilliporeSigma

Synonyms: 3,3-Bis(4-hydroxynaphthalen-1-yl)isobenzofuran-1(3H)-one

1.06246.0001 1.06246.0005
EM1.06246.0001CS 17685 USD
EM1.06246.0001 EM1.06246.0005
α-Naphtholphthalein indicator, Supelco®
α-Naphtholphthalein
Formula: C₂₈H₁₈O₄
MW: 418.45 g/mol
MDL Number: MFCD00036202
CAS Number: 596-01-0
EINECS: 209-875-5

Order Now

Specification Test Results

Solubility (20 °C) practically insoluble
Melting Point 253 - 255 °C
Molar Mass 418.45 g/mol
Bulk Density 350 kg/m3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR