N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Marshall's Reagent, N-2-Aminoethyl-1-naphthylamine dihydrochloride, N-1-Naphthylethylenediamine

1.06237.0005
EM1.06237.0005CS 5829 USD
EM1.06237.0005
N-(1-Naphthyl)ethylenediamine dihydrochloride, GR for analysis, Supelco®
N-2-Aminoethyl-1-naphthylamine dihydrochloride

1.06237.0025 replaces and is the same as NX0230-1. Part # Change only, Same Pack Size, Same Specifications.

Formula: C₁₂H₁₆Cl₂N₂
MW: 259.18 g/mol
Melting Pt: 188…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00012556
CAS Number: 1465-25-4
EINECS: 215-981-2
Merck Index: 12,06496
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3643e6df3e9cf8dc627823e9090c97004ee25308&vid=a30fc28279c11af12c6b735b00b84fc8f08caf34&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 30 g/l (20 °C)
Molar Mass 259.18 g/mol
Bulk Density 380 kg/m3
pH 1.0 (25 g/l, H2O, 20 °C)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR