Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ100

1.06107.0003
EM1.06107.0003CS 4888 USD
EM1.06107.0003
Molecular sieve 3A (0.3 nm, 3 Å) absorption tube for H₂O, with indicator, Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)

Absorption tube for H₂O (molecular sieve 0.3 nm with indicator)

Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
Storage Temperature: Ambient
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR