2-Methylbutane, Uvasol® for spectroscopy, Supelco®

Supplier: MilliporeSigma
Danger

1.06056.1000
EM1.06056.1000CS 1811 USD
EM1.06056.1000
2-Methylbutane, Uvasol® for spectroscopy, Supelco®
2-Methylbutane

Accurate analytic results in UV/VIS and infrared spectroscopy depend on the use of very pure solvents for sample preparation. The Uvasol® solvents range has been specially designed for spectroscopy and other applications requiring solvents of the highest spectral purity. The refinement process allows a greater degree of security in applications and avoids misinterpretation of analytical results caused by traces of UV, IR and fluorescence contamination. Uvasol® solvents offer best UV transmittance. In all specifications the minimum transmittance for 5 typical wavelengths are identified. Furthermore the transmittance is specified in accordance with Reag. Ph Eur and ACS.

Formula: CH₃CH₂CH(CH₃)₂
Boiling Pt: 28 °C (1013 hPa)
Melting Pt: –160 °C
Density: 0.62 g/cm³ (20 °C)
Flash Pt: –51 °C
Storage Temperature: Ambient
MDL Number: MFCD00009338
CAS Number: 78-78-4
EINECS: 201-142-8
UN: 1265
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=498904a9f8b2618e19d2343aa74e247ec7121f00&vid=c7ed2044255d0d72af23a3fafec4ef6066d25097&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 0.048 g/l (20 °C)
Melting Point -160 °C
Molar Mass 72.15 g/mol
Boiling Point 28 °C (1013 hPa)
Vapor Pressure 800 hPa (20 °C)
Flash Point -57 °C
Explosion Limit 1.3 - 7.6 %(V)
Density 0.62 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 420 °C
Evaporation number 1
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR