Dichloromethane ≥99.8% stabilised, SupraSolv® for gas chromatography, for chromatography, distilled, Supelco®

Supplier: MilliporeSigma
Warning


Stabilization: Stabilised with 2-methyl-2-butene (iso-amylene)

1.06054.1000 1.06054.2500 1.06054.4000
EM1.06054.1000CS 668 USD
EM1.06054.1000 EM1.06054.2500 EM1.06054.4000
Dichloromethane ≥99.8% stabilised, SupraSolv® for gas chromatography, for chromatography, distilled, Supelco®
Dichloromethane

SupraSolv® ECD and FID is specially developed for gas chromatography in combination with ECD (Electron Capture Detetor) and FID (Flame Ionization detector). SupraSolv® solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results. Typical applications include the determination of polychlorinated biphenyls (PCB) in water and soil or pesticides in fruits and vegetables. Also ideal for many EPA protocols.

Formula: CH₂Cl₂
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1,III
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity (GC) =99.8%
Identity(IR) Conforms
Evaporation Residue = 5.0 mg/l
Water = 0.01%
Color =10 Hazen
GC/ECD (retention range 1,2,4-trichlorobenzene to decachlorobiphenyle, individual signals (lindane standard)) = 3pg/ml
GC/FID (retention range n-undecane to n-tetracontane, individual signals (n-tetradecane standard)) = 3ng/ml
Suitability Residue Analysis


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR