Dichloromethane, LiChrosolv® for liquid chromatography, Supelco®
Supplier: MilliporeSigma
|
Warning
|
M-Clarity™ quality level = MQ100
1.06044.1000
1.06044.2500
1.06044.4000
EM1.06044.1000CS
632
USD
EM1.06044.1000
EM1.06044.2500
EM1.06044.4000
Dichloromethane, LiChrosolv® for liquid chromatography, Supelco®
Dichloromethane
With their high degree of UV transmittance, low particle count, low acidity and alkalinity and low evaporation residue level, LiChrosolv® solvents are ideal for reproducible separations. Since separations are normally carried out under gradient conditions in analytical HPLC, we offer solvents in "gradient grade" as well as "isocratic grade". This enables to minimize the gradient effect of the solvent involved.
Formula:
CH₂Cl₂ Boiling Pt: 39.8 °C (1013 hPa) Melting Pt: –95 °C Density: 1.322 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000881 CAS Number: 75-09-2 EINECS: 200-838-9 UN: 1593 ADR: 6.1,III Merck Index: 12,06140 |
|
Specification Test Results
Solubility | 20 g/l (20 °C) |
Melting Point | -95 °C |
Molar Mass | 84.93 g/mol |
Boiling Point | 40 °C (1013 hPa) |
Vapor Pressure | 475 hPa (20 °C) |
Refractive Index | 1.42 |
Explosion Limit | 13 - 22 %(V) |
Density | 1.33 g/cm3 (20 °C) |
pH | (H2O, 20 °C) neutral |
Ignition Point | 605 °C DIN 51794 |
Synonyms | DCM, Methylene chloride, Methylene dichloride |
Hill Formula | CH2Cl2 |
Chemical Formula | CH2Cl2 |
LD50 oral | LD50 Rat 1600 mg/kg |
LD50 dermal | LD50 Rat greater than 2000 mg/kg |
Storage | Store at +15°C to +25°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...