Methanol, anhydrous (max. 0.003% H₂O), Seccosolv®, Supelco®

Supplier: MilliporeSigma
Danger

1.06012.1000 1.06012.2500
EM1.06012.0500CS 164.76 USD
EM1.06012.0500 EM1.06012.1000 EM1.06012.2500 EM1.06012.6010
Methanol, anhydrous (max. 0.003% H₂O), Seccosolv®, Supelco®
Methanol

These reagents are produced using specially selected distillation methods that ensure consistently high dryness and batch-to-batch consistency. To protect the quality of these products even better from potential contaminants, a septum seal cap provides multiple layers of protection to keep solvents in flawless condition before, during, and after removal.

Formula: CH₃OH
Boiling Pt: 64.6 °C (1013 hPa)
Melting Pt: –98 °C
Density: 0.7918 g/cm³ (20 °C)
Flash Pt: 11 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004595
CAS Number: 67-56-1
EINECS: 200-659-6
UN: 1230
ADR: 3 (6.1),II
REACH: 01-2119433307-44
Merck Index: 13,05984
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ea983428c5015ad8417c1e1c260cce3a4d1de3c&vid=9bc2f21fdce2dcabce477372374ff56b8fb3a0f1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility (20 °C) soluble
Melting Point -98 °C
Molar Mass 32.04 g/mol
Boiling Point 64.5 °C (1013 hPa)
Vapor Pressure 128 hPa (20 °C)
Flash Point 10 °C
Refractive Index 1.33
Explosion Limit 5.5 - 44 %(V)
Density 0.792 g/cm3 (20 °C)
pH (H2O) no data available
Ignition Point 455 °C DIN 51794
Evaporation number 6.3
Water absorption 1000 g/kg
Hazard Class 3
Hazard Class Description Flammable liquid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR