D-(-)-Mannitol for determination of boric acid, Supelco®

Supplier: MilliporeSigma

Synonyms: Mannitol, (2R,3R,4R,5R)-Hexane-1,2,3,4,5,6-hexaol

1.05983.0250 1.05983.1000
EM1.05983.0250CS 1216 USD
EM1.05983.0250 EM1.05983.1000
D-(-)-Mannitol for determination of boric acid, Supelco®
D(-)-Mannitol
Formula: HOH₂C(CH(OH))₄CH₂OH
MW: 182.17 g/mol
Boiling Pt: 290…295 °C (4 hPa)
Melting Pt: 164…169 °C
Density: 1.49 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064287
CAS Number: 69-65-8
EINECS: 200-711-8
Merck Index: 13,05769
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=163f556dca45086e05ff490ddc8eeb65d04aea05&vid=408e3f5566e309064ce68988cdd160305c5b8c0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Solubility 213 g/l (25 °C)
Melting Point 164 - 169 °C
Molar Mass 182.17 g/mol
Bulk Density 400 - 500 kg/m3
Boiling Point 290 - 295 °C (4 hPa)
Vapor Pressure (20 °C)
Density 1.49 g/cm3 (20 °C)
pH 5 - 7 (100 g/l, H2O, 20 °C)
Ignition Point 410 °C
Viscosity kinematic (20 °C) not applicable


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR