Manganese(IV) oxide, powder, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

Synonyms: Manganese dioxide, Manganese(IV) oxide, activated, Manganese peroxide, Mangan(IV)-oxid

1.05957.1000
EM1.05957.1000CS 1619 USD
EM1.05957.1000
Manganese(IV) oxide, powder, EMSURE® for analysis, Supelco®
Manganese (IV) oxide
Formula: MnO₂
MW: 86.94 g/mol
MDL Number: MFCD00003463
CAS Number: 1313-13-9

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Molar Mass 86.94 g/mol
Bulk Density 600 - 800 kg/m3
Vapor Pressure (20 °C)
Density 5.03 g/cm3 (20 °C)
pH (H2O) not applicable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR