Magnesium sulfate heptahydrate 99.0-100.5% (by complexometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®

Supplier: MilliporeSigma

Synonyms: sulphuric acid magnesium salt heptahydrate, sulfuric acid magnesium salt heptahydrate, Epsom salt, Bitter salt

1.05882.9029
EM1.05882.2500CS 624 USD
EM1.05882.2500 EM1.05882.9029
Magnesium sulfate heptahydrate 99.0-100.5% (by complexometric analysis, on dry substance), EMPROVE® ESSENTIAL Ph. Eur., BP, USP, SAFC®
Magnesium sulphate heptahydrate
Formula: MgSO₄·7H₂O
MW: 246.48 g/mol
Density: 1.68 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149785
CAS Number: 10034-99-8
EINECS: 231-298-2

Order Now

Specification Test Results

Assay (complexometric; calculated on dried substance) 99.0 - 100.5 %
Identity passes test
Appearance of solution passes test
pH-value (5?%; water) 5.0 - 9.2
Acidity or alkalinity conforms
Chloride (Cl) ≤ 0.014%
As (Arsenic) ≤ 0.0002 %
Fe (Iron) ≤ 0.001 %
Se (Selenium) ≤ 0.001 %
Loss on drying (450?°C) 49.0 - 52.0 %
Residual solvents (ICH Q3C) Excluded by production process
Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).
Corresponds to Ph Eur, BP, USP

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR