Magnesium nitrate hexahydrate 99.0-102.0% (by complexometric analysis), EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Warning
|
1.05853.0500
1.05853.9025
EM1.05853.0500CS
1426
USD
EM1.05853.0500
EM1.05853.9025
Magnesium nitrate hexahydrate 99.0-102.0% (by complexometric analysis), EMSURE® for analysis, Supelco®
Magnesium nitrate hexahydrate
Formula:
Mg(NO₃)₂·6H₂O Boiling Pt: 330 °C (1013 hPa) Melting Pt: 89 °C (for the hexahydrate) Density: 1.46 g/cm3 (20 °C) Flash Pt: 93.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00149779 CAS Number: 13446-18-9 EINECS: 233-826-7 UN: 1474 ADR: 5.1,III Merck Index: 13,05697 |
Specification Test Results
Solubility | 420 g/l (20 °C) |
Melting Point | 89 °C (for the hexahydrate) |
Molar Mass | 256.41 g/mol |
Density | 1.46 g/cm3 (20 °C) |
pH | 5.0 - 7.0 (50 g/l, H2O, 25 °C) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...