Magnesium, powder ∼ 0.06-0.3 mm, Supelco®

Supplier: MilliporeSigma
Danger

1.05815.1000
EM1.05815.1000CS 2075 USD
EM1.05815.1000
Magnesium, powder ∼ 0.06-0.3 mm, Supelco®
Magnesium
Formula: Mg
MW: 24.3 g/mol
MDL Number: MFCD00085308
CAS Number: 7439-95-4
UN: 1418
ADR: 4.3 (4.2),II

Order Now

Specification Test Results

Solubility (20 °C) (slow decomposition)
Melting Point 651 °C
Molar Mass 24.31 g/mol
Boiling Point 1107 °C (1013 hPa)
Vapor Pressure 0.00013 hPa (325 °C)
Density 1.74 g/cm3 (20 °C)
Ignition Point 450 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR