Magnesium nitrate (≥10.0 ± 0.2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®

Supplier: MilliporeSigma
Danger


M-Clarity™ quality level = MQ100

1.05813.0050
EM1.05813.0050CS 6776 USD
EM1.05813.0050
Magnesium nitrate (≥10.0 ± 0.2 g/L Mg) in nitric acid 17% matrix modifier solution, for graphite furnace AAS, Supelco®
Magnesium nitrate solutions

Magnesium matrix modifier for graphite furnace AAS.

Formula: Mg(NO₃)₂
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
CAS Number: 10377-60-3
UN: 2031
ADR: 8,II

Order Now

Specification Test Results

Ag (Silver) ≤ 0.000002 g/l
As (Arsenic) ≤ 0.00001 g/l
Cd (Cadmium) ≤ 0.000001 g/l
Pb (Lead) ≤ 0.00001 g/l
Se (Selenium) ≤ 0.0001 g/l
Tl (Thallium) ≤ 0.00001 g/l

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR