Magnesium, foil, Width 3 mm, Thickness 0.15-0.30 mm, Supelco®

Supplier: MilliporeSigma
Warning

1.05812.0001
EM1.05812.0001CS 110725 USD
EM1.05812.0001
Magnesium, foil, Width 3 mm, Thickness 0.15-0.30 mm, Supelco®
Magnesium
Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107
Melting Pt: 651
MDL Number: MFCD00085308
CAS Number: 7439-95-4
UN: 1869
ADR: 4.1 III
Merck Index: 13,05675

Order Now

Specification Test Results

Melting Point 651 °C
Molar Mass 24.31 g/mol
Boiling Point 1107 °C
Density 1.75 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR