TLC Plates, Silica Gel 60 F254 Coated, MilliporeSigma
Supplier: MilliporeSigma
1.05794.0001
EM1.05794.0001EA 297.25 USD
EM1.05794.0001
TLC Plates, Silica Gel 60 F254 Coated, MilliporeSigma
TLC Plates
These TLC Plates are coated in TLC silica gel 60 F254
Product # 1.05794.0001 replaces and is identical to Product # 15341-1.
They are glass-backed and cover almost every type of separation when combined with a suitable mobile phase.