Molecular sieve A4 (0.4 nm, 4 Å), rods ~ 1.6 mm (1/16"), Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.05743.1000
EM1.05743.1000CS 2536 USD
EM1.05743.1000
Molecular sieve A4 (0.4 nm, 4 Å), rods ~ 1.6 mm (1/16"), Supelco®
Density: 1.1…2.2 g/cm³ (20 °C)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR