Molecular sieve 3A (0.3 nm, 3 Å), rods ~ 1.6 mm (1/16"), Supelco®

Supplier: MilliporeSigma

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

1.05741.0250
EM1.05741.0250CS 2346 USD
EM1.05741.0250
Molecular sieve 3A (0.3 nm, 3 Å), rods ~ 1.6 mm (1/16"), Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Bulk Density 700 - 750 kg/m3
Synonyms Sodium aluminium silicate

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR