Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm / ~ 10 mesh, with indicator, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ200

1.05739.0250
EM1.05739.0250CS 518 USD
EM1.05739.0250
Molecular sieve A4 (0.4 nm, 4 Å), beads ~ 2 mm / ~ 10 mesh, with indicator, Supelco®
Molecular sieve A4 (0.4 nm, 4 Å)
Density: 1.1…2.2 g/cm³ (20 °C)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR