Molecular sieve 3A (0.3 nm, 3 Å), Supelco®

Supplier: MilliporeSigma

Synonyms: Molecular sieve 0.3 nm, 3A molecular sieve, Molecular sieve A3, Linde 3A, Sodium aluminium silicate

M-Clarity™ quality level = MQ200

1.05734.1000
EM1.05734.1000CS 1679 USD
EM1.05734.1000
Molecular sieve 3A (0.3 nm, 3 Å), Supelco®
Molecular sieve 3A (0.3 nm, 3 Å)
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR