Molecular sieve 3A (0.3 nm, 3 Å), beads ~ 2 mm, with moisture indicator, Supelco®

Supplier: MilliporeSigma

1.05734.0250
EM1.05734.0250CS 2346 USD
EM1.05734.0250
Molecular sieve 3A (0.3 nm, 3 Å), beads ~ 2 mm, with moisture indicator, Supelco®
Formula: KnNa₁₂-n[(AlO₂)₁₂(SiO₂)₁₂]·xH₂O
MDL Number: MFCD00131613
CAS Number: 308080-99-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR