Molecular sieve A4 (0.4 nm, 4 Å), beads Reag. Ph. Eur. ~ 2 mm, Supelco®

Supplier: MilliporeSigma

1.05708.0250 1.05708.1000
EM1.05708.0250CS 855 USD
EM1.05708.0250 EM1.05708.1000
Molecular sieve A4 (0.4 nm, 4 Å), beads Reag. Ph. Eur. ~ 2 mm, Supelco®
Molecular sieve A4 (0.4 nm, 4 Å)
MDL Number: MFCD03457537
CAS Number: 70955-01-0

Order Now

Specification Test Results

Solubility (20 °C) insoluble
Bulk Density 700 - 750 kg/m3
Grade Reag. Ph Eur
Synonyms Sodium aluminium silicate


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR