Lithium carbonate, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.05680.0250
EM1.05680.0250CS 4990 USD
EM1.05680.0250
Lithium carbonate, EMSURE® for analysis, Supelco®
Lithium carbonate

1.05680.0250 replaces and is the same as LX0325-1. Different Pack Size, Same Specifications.
1.05680.0250 is offered as an alternative to LX0325-20. different size, different packaging, same chem spec.

Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720
Density: 2.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Specification Test Results

Solubility 13 g/l (20 °C)
Melting Point 720 °C
Molar Mass 73.89 g/mol
Bulk Density 250 kg/m3
Vapor Pressure (20 °C)
Density 2.1 g/cm3 (20 °C)
pH 10 - 11 (5 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR