Lithium carbonate ≥98.5% (by acidimetry), Supelco®

Supplier: MilliporeSigma
Warning


M-Clarity™ quality level = MQ200

1.05670.9050
EM1.05670.9050EA 17324 USD
EM1.05670.9050
Lithium carbonate ≥98.5% (by acidimetry), Supelco®
Lithium carbonate
Formula: Li₂CO₃
MW: 73.89 g/mol
Melting Pt: 720 °C
Density: 2.1 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011084
CAS Number: 554-13-2
EINECS: 209-062-5
Merck Index: 13,05549

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR