Sodium silicate solution (7.5 - 8.5% Na₂O; 25.5 - 28.5% SiO₂), extra pure, Supelco®

Supplier: MilliporeSigma
Danger

1.05621.2500 1.05621.9040 1.05621.9250
EM1.05621.2500CS 778 USD
EM1.05621.2500 EM1.05621.9040 EM1.05621.9250
Sodium silicate solution (7.5 - 8.5% Na₂O; 25.5 - 28.5% SiO₂), extra pure, Supelco®
Sodium silicate
Formula: Na₂O(SiO₂)x·xH₂O
Boiling Pt: >100 °C (1013 hPa)
Density: 1.35 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003492
CAS Number: 1344-09-8
UN: 1719
ADR: 8,II

Order Now

Specification Test Results

Identity passes test
Assay (acidimetric, Na₂O) 7.5 - 8.5 %
Assay (acidimetric, SiO₂) 25.5 - 28.5 %
Density (20 °C) 1.296 - 1.396 g/ml
Carbonate (as CO₂) passes test
Fe (Iron) ≤ 50 ppm
Heavy metals (as Pb) ≤ 50 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR