Sodium hydroxide ∼32% in aqueous solution, EMSURE® for nitrogen analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

1.05590.2500 1.05590.9025
EM1.05590.2500CS 606 USD
EM1.05590.2500 EM1.05590.9025
Sodium hydroxide ∼32% in aqueous solution, EMSURE® for nitrogen analysis, Supelco®
Sodium hydroxide
Formula: NaOH
Boiling Pt: 119 - 135 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.35 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Assay (acidimetric,NaOH) ≥ 32.0 %
Carbonate (as Na₂CO₃) ≤ 0.2 %
Chloride (Cl) ≤ 0.004 %
Phosphate (PO₄) ≤ 0.0005 %
Silicate (SiO₂) ≤ 0.001 %
Sulfate (SO₄) ≤ 0.003 %
Total nitrogen (N) ≤ 0.0001 %
Heavy metals (as Pb) ≤ 0.0005 %
Al (Aluminium) ≤ 0.0005 %
Fe (Iron) ≤ 0.0005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR