Sodium hydroxide ≥10% in aqueous solution, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

1.05588.1000 1.05588.9010
EM1.05588.1000CS 383 USD
EM1.05588.1000 EM1.05588.9010
Sodium hydroxide ≥10% in aqueous solution, EMSURE® for analysis, Supelco®
Sodium hydroxide

Basic cleaning agent (concentration ~10%) in premium MilliporeSigma quality. The high purity of these solutions make these sodium liquids suitable above all for cleaning applications in which residues from surfactants or complexation agents are to be avoided.


  • Strongly alkaline liquid
  • Chlorine-free


Through the use of these prepared solutions, the time-consuming and dangerous breakdown of solid sodium hydroxide can be avoided.

Formula: NaOH
Boiling Pt: 119…135 °C (1013 hPa)
Melting Pt: 9…10 °C
Density: 1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.11 g/cm3 (20 °C)
pH 13.8 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR