Iron(III) chloride (10% Fe) in aqueous solution, EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Iron trichloride, Ferric chloride

1.05512.0250
EM1.05512.0250CS 2126 USD
EM1.05512.0250
Iron(III) chloride (10% Fe) in aqueous solution, EMSURE® for analysis, Supelco®
Iron (III) chloride
Formula: FeCl₃
MDL Number: MFCD00011005
CAS Number: 7705-08-0
EINECS: 231-729-4
UN: 2582
ADR: 8,III

Order Now

Specification Test Results

Solubility (20 °C) soluble
Density 1.29 g/cm3 (20 °C)
pH  - 1 (H2O, 20 °C)
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR