Ammonium sulfide ≥20% in aqueous solution, GR analytical reagent, Supelco®

Supplier: MilliporeSigma
Danger

1.05442.1000
EM1.05442.1000CS 2611 USD
EM1.05442.1000
Ammonium sulfide ≥20% in aqueous solution, GR analytical reagent, Supelco®
Ammonium sulphide
Formula: H₈N₂S
MW: 68.14 g/mol
MDL Number: MFCD00010892
CAS Number: 12135-76-1
EINECS: 235-223-4
UN: 2683
ADR: 8 (3, 6.1),II

Order Now

Specification Test Results

Solubility (20 °C) soluble
Vapor Pressure 450 hPa (20 °C)
Flash Point 15 °C
Explosion Limit 4 - 46 %(V)
Density 0.997 g/cm3 (20 °C)
pH 9.5 (200 g/l, H2O, 25 °C)
Hazard Class 8
Hazard Class Description Corrosive material

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR