Potassium sulfate, Supelco®

Supplier: MilliporeSigma


M-Clarity™ quality level = MQ300

1.05153.9025
EM1.05153.9025EA 2156.56 USD
EM1.05153.9025
Potassium sulfate, Supelco®
Potassium sulphate
Formula: K₂SO₄
MW: 174.26 g/mol
Boiling Pt: 1698
Melting Pt: 1069
Density: 2.66 g/cm³ (20 °C)
MDL Number: MFCD00011388
CAS Number: 7778-80-5
Merck Index: 13,07759

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR