Potassium sorbate 99.0-101.0%, granules, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E202, SAFC®

Supplier: MilliporeSigma
Warning

Synonyms: Potassium 2,4-Hexadienoate, Sorbic acid potassium salt, Potassium (2E,4E)-hexa-2,4-dienoate

1.05119.9025
EM1.05119.9025EA 4016.63 USD
EM1.05119.9025
Potassium sorbate 99.0-101.0%, granules, EMPROVE® ESSENTIAL Ph. Eur., BP, NF, FCC, E202, SAFC®
Potassium sorbate

"Residues of metal catalysts or metal reagents acc. to EMEA/CHMP/SWP/4446/2000 are not likely to be present. Conforms to the purity criteria on food additives according to the current European Commission Regulation. Corresponds to Ph Eur, BP, NF, FCC, E 202"

Formula: CH₃CH=CHCH=CHCOOK
MW: 150.22 g/mol
Melting Pt: 270 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00016546
CAS Number: 24634-61-5
EINECS: 246-376-1
Merck Index: 13,07756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=500421971592bd8f37c1a6b88f97c1de033cbf87&vid=e8c4a1d0da78946f174af6a94e5c78fc9c16de05&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (acidimetric; calculated on dried substance) 99.0 - 101.0 %
Identity passes test
Appearance of solution (5 %; water) passes test
Acidity or alkalinity passes test
Heavy metals (as Pb) ≤ 0.0010 %
As (Arsenic) ≤ 0.0003 %
Cd (Cadmium) ≤ 0.001 %
Cu (Copper) ≤ 0.001 %
Hg (Mercury) ≤ 0.0001 %
Pb (Lead) ≤ 0.0002 %
Zn (Zinc) ≤ 0.001 %
Aldehydes (as Acetaldehyd) ≤ 0.15 %
Residual solvents (Ph.Eur./USP/ICH) class 3 ≤ 0.5 %
Other residual solvents (Ph.Eur./USP/ICH) excluded by production process
Thermal stability (105 °C) passes test
Loss on Drying (105°C) ≤ 1.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR