Potassium fluoride, EMSURE® for analysis, Supelco®
Supplier: MilliporeSigma
|
Danger
|
1.04994.0250
1.04994.1000
EM1.04994.0250CS
1146.1
USD
EM1.04994.0250
EM1.04994.1000
Potassium fluoride, EMSURE® for analysis, Supelco®
Potassium fluoride
Formula:
KF Boiling Pt: 1505 °C (1013 hPa) Melting Pt: 860 °C Density: 2.48 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011398 CAS Number: 7789-23-3 EINECS: 232-151-5 UN: 1812 ADR: 6.1,III Merck Index: 13,07714 |
Specification Test Results
Solubility | 923 g/l (20 °C) |
Melting Point | 860 °C |
Molar Mass | 58.1 g/mol |
Bulk Density | 400 kg/m3 |
Boiling Point | 1500 °C (1013 hPa) |
Vapor Pressure | 1.3 hPa (885 °C) |
Density | 2.48 g/cm3 (20 °C) |
pH | 7 - 9 (50 g/l, H2O, 18 °C) |
Hazard Class | 6.1 |
Hazard Class Description | Toxic Materials |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...