Potassium hexacyanoferrate(III), EMSURE® for analysis, Supelco®

Supplier: MilliporeSigma
Warning

1.04973.0100 1.04973.0250 1.04973.1000 1.04973.9050
EM1.04973.0100CS 8446 USD
EM1.04973.0100 EM1.04973.0250 EM1.04973.1000 EM1.04973.9050
Potassium hexacyanoferrate(III), EMSURE® for analysis, Supelco®
Potassium hexacyanoferrate (III)
Formula: K₃Fe(CN)₆
Density: 1.85 g/cm3 at 20 °C
Storage Temperature: Ambient
MDL Number: MFCD00011392
CAS Number: 13746-66-2
EINECS: 237-323-3
UN: 3077
ADR: 9,III
Merck Index: 13,07712

Order Now

Specification Test Results

Solubility 464 g/l (20 °C)
Molar Mass 329.25 g/mol
Bulk Density 900 - 1000 kg/m3
Density 1.85 g/cm3 (20 °C)
pH 6 (50 g/l, H2O, 20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR